Would you like to log in again?
MANIFOLD SYSTEM M30 30-50L
Supplier: Chemglass
High Flow Manifold System with Hard Connect Fittings^ 30L-50L Reactor Capacity, M30 Circulator Size End Fitting, 1in Beaded Pipe Triple Insulated Flexible Hose, -60 to 200 degC Temperature Range. System is supplied with 3/4in ID Triple Insulated Flexible Hose.
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
MANIFOLD SYSTEM M30 30-50L 89028-896 CG-1969-M-06 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty