Would you like to log in again?
ELLIPSO-SPOON 28 X 65 X 300MM
Supplier: Chemglass
Ellispso-Spoon* Spatula^ Stainless Steel. Spoon Size - 28 x 65, Overall Length - 300mm, handle dia - 8mm. Specialty curved shape. Crystals, powders, solid substances and thick liquids are easily retrieved and do not fall back into the container.
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
ELLIPSO-SPOON 28 X 65 X 300MM 89028-938 CG-1987-03 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty