Would you like to log in again?
STIR BAR MAGNETIC 50X12MM
Supplier: Chemglass
Triangular, Magnetic Stir Bar^ PTFE, 50mm L X 12mm Diameter. Effective for dissolving solids and mixing sediments in a flat bottom container such as a beaker or erlenmeyer flask.
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
STIR BAR MAGNETIC 50X12MM 89028-988 CG-2009-16 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty