Would you like to log in again?
STIR BAR MAGNETIC X 20X8MM
Supplier: Chemglass
Magnetic X Shaped Stir Bar^ PTFE, 20mm Diameter X 8mm Height. Shape of stir bar creates a deep vortex and provides stability during stirring.
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
STIR BAR MAGNETIC X 20X8MM 89028-996 CG-2013-02 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty