Would you like to log in again?
STOPPER TEFLON 19/22 WITH
Supplier: Chemglass
19/22 PTFE* Stopper^ Stopper has a Polypropylene extraction nut permitting easy removal of the stopper from the glass outer joint.
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
STOPPER TEFLON 19/22 WITH 89029-060 CG-3002-L-19 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty