Would you like to log in again?
Lattice Support Shelf, Chemglass
Supplier: Chemglass
CG-3050-10
89029-162EA 283.88 USD InStock
89029-162
Lattice Support Shelf, Chemglass
Support Stands
This shelf can be attached to any lattice setup clamp holder.
  • Anodized green with a neoprene surface to prevent from slipping off
The ½" square support rod is 7.5" long. Square support rod prevents shelf from rotating once it is attached to the lattice.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty