Brown, low compression, 75 durometer Viton® O-rings are suitable for acids, aliphatic hydrocarbons, oxidizing agents, and all petroleum products
Note: Viton® O-ring 89029-223 is not a standard size.
They are especially good for vacuum service and low gas permeability. Temperature range: –30 to 200°C (–22 to 392°F).