Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
Double-Bank Vacuum/Inert Manifold, Chemglass
Supplier: Chemglass
CG-4440-01 CG-4440-02 CG-4441-03 CG-4441-02 CG-4441-01 CG-4440-03
89029-404EA 1417.16 USD InStock
89029-404 89029-406 89029-408 80072-476 80072-478 80072-474
Double-Bank Vacuum/Inert Manifold, Chemglass
Manifolds
General purpose, double-bank manifold for use with vacuum and inert gas.
Both banks are made from 1/2" medium wall tubing with 10 mm (0.39") O.D. serrated connections. Stopcocks are glass 4 mm (0.16") bore with lower vacuum chamber and are spaced 85 mm (3.35") center to center. Manifold has top right connection to inert gas and lower left connection for vacuum.
For vacuum manifold model both ends of inert gas line open to permit connection to the gas source and an oil bubbler.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty