Would you like to log in again?
Sample Vials, Amber, PTFE Lined Caps, Chemglass
Supplier: Chemglass
CG-4905-03 CG-4905-02 CG-4905-06 CG-4905-04 CG-4905-05 CG-4905-01
89029-458EA 487.29 USD InStock
89029-458 89029-460 89029-462 89029-464 89029-466 89029-468
Sample Vials, Amber, PTFE Lined Caps, Chemglass
Vials
Sample vials are made from Type 1 Borosilicate glass.
  • Full height partitions minimize breakage
  • Solid PTFE lined caps
Packaging: Packaged in convenient LAB-PACS of 144 pieces.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty