Would you like to log in again?
Crucibles with Lids, Quartz, Chemglass
Supplier: Chemglass
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
CGQ-4000-03 CGQ-4000-01 CGQ-4000-02 CGQ-4000-04
89029-680EA 200.71 USD InStock
89029-680 89029-682 89029-684 89029-686
Crucibles with Lids, Quartz, Chemglass
Crucibles
Crucibles are suitable for use in temperatures up to 1000°C, which is perfect for use in muffle furnace ashing procedures.
  • Made of quartz
  • Withstands temperatures up to 1000°C
Quartz material ensures sample purity for further analysis.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty