Would you like to log in again?
Flexible Graphite Gasket, Parr Instrument Company
Supplier: Parr Instrument Company
457HC3KL
89029-764PK 64.89 USD InStock
89029-764
Flexible Graphite Gasket, Parr Instrument Company
Seals Gaskets
A torque wrench should be used to tighten the compression bolts on vessels equipped with a flexible graphite gasket. Since used on various Parr high pressure vessels, the user must be sure that the torque wrench is fitted with an adapter which matches the bolts on which it will be used.
It may be possible to extend the useful life of a flexible graphite gasket by coating the sealing surfaces on both the vessel and the gasket with a silicone-base lubricant.
Max. pressure: 6000 psi
Torque: 40 feet-lbs.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty