Would you like to log in again?
SP Bel-Art Spinbar® Magnetic Stirring Bar Garage, Bel-Art Products, a part of SP
Supplier: Bel-Art
Spinbar®
F37790-0000
89029-962EA 144.19 USD InStock
89029-962
SP Bel-Art Spinbar® Magnetic Stirring Bar Garage, Bel-Art Products, a part of SP
Stirrers Magnetic Stirring Bars
Keep all magnetic stirring bars and accessories organized and accessible with the Spinbar® Magnetic Stirring Bar Garage.
  • Stirring bars easily demagnetized
  • Epoxy coating prolongs life and allows easy cleaning
  • Rack may be wall-mounted or free-standing
  • Multiple wire shelves
Multiple sizes and shapes of magnetic stirring bars from large 15.9 cm (6”) to tiny 10 mm (0.5”) bars can be stored on this rack.
Stirring bars are easily demagnetized when stored together in a box or drawer. The best way to store them is in pairs, with the north pole of one adjoining the south pole of the other. Tight wire spacing on the upper section of the rack holds small bars, while wider spacing below can be used for larger bars. A lower wire shelf is ideal for holding small lab tools including the Spinbar® Restrainer.
Epoxy-coated for long life and easy cleaning, the rack is freestanding or can be mounted to a vertical surface. Use one rack to dry stirring bars after cleaning, and place others near associated stirrers to keep bars organized and visible.
Ordering information: Mounting hardware not included.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty