Would you like to log in again?
VWR® Reclosable Pink Anti-Static Bags
Supplier: VWR
VWR®
MGST4P0406 MGST4P1318 MGST4P0305 MGST4P0912 MGST4P0608
89005-348CS 82.36 USD InStock
89005-348 89005-352 89005-350 89030-074 89030-072
VWR® Reclosable Pink Anti-Static Bags
Bags Sample Bags
These flexible and electrostatic-free 4 mm bags qualify as Type II entitled barrier material under MIL-B-81750b.
  • Amine-free
  • Reclosable design
  • Pink tint for easy identification
They dissipate an induced 5000 V charge to technical zero in accordance with FTS 101C, Method 4046.1. Bags protect contents from damage caused by static electricity, in addition to providing protection against dust and moisture.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty