Would you like to log in again?
Amplitude™ EcoCloth™ Wipes, Contec®
Supplier: Contec
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
Amplitude™ EcoCloth™
AMEC0003 AMEC0005 AMEC0001 AMEC0004 AMEC0002
89030-168CS 358.19 USD InStock
89030-168 89030-166 89030-164 89030-172 89030-170
Amplitude™ EcoCloth™ Wipes, Contec®
Wipes Cleanroom Wipes Dry Cleanroom Wipes
The EcoCloth™ wipe has been engineered to provide greater thickness in a lighter weight material. Amplitude™ EcoCloth™ is a hydroentangled cellulose/polyester fabric with additional loft. It is lightweight but has excellent sorbency which exceeds comparable standard cellulose/polyester wipes.
  • Recommended for ISO Class 5/6 environments
  • Low basis weight cellulose/polyester wipe with additional loft for greater sorbency
  • Most sorbent of all cellulose/polyester wipes
  • Lighter weight with the same performance, wipes can reduce solid waste in excess of 40%
  • Double packaged to preserve product integrity
The increased thickness greatly enhances the sorbent capacity and also makes it easier for operators to pick up one wipe at a time. Amplitude™ EcoCloth™ wipes are a cost-effective solution for general-purpose applications such as product transfer, wiping to dry, and absorbing spills.
When customers are looking for ways to reduce their solid waste disposal for either financial or environmental reasons, the combined effect of superior performance and lighter weight provides a potential reduction in solid waste in excess of 40% compared to standard cellulose/polyester wipes. The result is a very environmentally friendly choice.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty