Would you like to log in again?
Amplitude™ Theta™ Wipes, Contec®
Supplier: Contec
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
Amplitude™ Theta™
AMTH0001 AMTH0002
89030-190CS 337.24 USD InStock
89030-190 89030-192
Amplitude™ Theta™ Wipes, Contec®
Wipes Cleanroom Wipes Dry Cleanroom Wipes
Amplitude™ Theta™ Wipes are made from a lightweight hydroentangled cellulose/polyester, which creates an economical absorbent wipe ideal for general cleaning applications.
  • Recommended for ISO Class 6 to 8 environments
  • Economical and cost-effective nonwoven wipe
  • Lightweight sorbent wipe
  • Double packaged to preserve product integrity
These wipes offer excellent durability and sorbency. They are designed for nonabrasive applications, such as general-purpose wiping and polishing of surfaces.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty