Would you like to log in again?
VWR THERMOMETER WATERPROOF
Supplier: VWR International
VWR* Waterproof Digital Thermometer^On/Off Switch And Auto Power Off After 10 Minutes, Waterproof Design, Data Hold, Max/Min Memory, Extra Large Display, 125Mm Stainless Steel Probe, Plastic Probe Cover With Pocket Clip, Ce Mark Certified, -50/150C (-58/302F), Battery Included (1 Lr44)
ORDER
Expand All / Collapse All
Description VWR Catalog Number Unit Price Quantity
VWR THERMOMETER WATERPROOF 89030-418 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty