Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
VWR® Magnetic Rare Earth Stir Bars
Supplier: VWR
VWR®
001.2670.RE 001.2650.RE 001.2615.RE 001.2610.RE 001.2625.RE
89030-558PK 1075.79 USD InStock
89030-558 89030-556 89030-554 89030-552 89030-550
VWR® Magnetic Rare Earth Stir Bars
Stirrers Magnetic Stirring Bars
Molded in pure, FDA-approved PTFE, these samarium-cobalt elliptical magnets feature increased magnetic strength and are almost totally resistant to demagnetization.
  • Autoclavable
  • PTFE construction
  • Highly resistant to demagnetization
Rare Earth magnets are identified by a chemically inert carbon black spot.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty