Would you like to log in again?
Cimarec™ i Poly 15, Thermo Scientific
Supplier: Thermo Scientific
50094596 50094596
89030-746EA 3118.03 USD InStock
CA89030-746 89030-746
Cimarec™ i Poly 15, Thermo Scientific
Stirrers Magnetic Stirrers, Multi-position
Stirrers are 100% maintenance and wear free, in a fully synchronized multipoint platform ideal to conduct stirring applications.
  • Large, flat easy to clean work surfaces
  • Precise microprocessor control with bright digital display
  • Gradual start acceleration is gentle and ensures good magnetic coupling
  • Inductive drive ensures completely synchronized stirring at all positions
  • Operating conditions are -10 to 40 °C at 95% RH
  • Rugged powder coated stainless-steel housing
  • IP32 protection rated and CE marked
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty