Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Cimarec™ i Telesystem Multi-position Magnetic Stirrers, Thermo Scientific

Supplier: Thermo Scientific
50088135 50088135 50088077 50088009 50088009 50088077
89030-756EA 6558.39 USD InStock
89030-756 89030-744 CA89030-752 89030-752 CA89030-756 CA89030-744
Cimarec™ i Telesystem Multi-position Magnetic Stirrers, Thermo Scientific
Stirrers Magnetic Stirrers, Multi-position
  • Cimarec™ i Telesystem: Remote control by means of separate TELEMODUL control unit
  • Can be used at -10 to +56°C (100% relative humidity), in an incubator
  • Immersible stirrers are ideal for use directly in water baths up to +50°C
  • Simultaneous control of multiple units with synchronised speed and identical operating conditions can be achieved with optional benchtop distributor

There are units with up to 60 stirring positions with TELEMODUL 20 °C controller has four power settings: 25, 50, 75 and 100% and can store three programmes and a RS232 interface.

Certifications: IP protection class according to DIN EN 60529: IP 68

Ordering information: Supplied with control units.

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty