Would you like to log in again?
SK Cleanroom Nitrile Gloves, Honeywell Safety
Supplier: Honeywell Safety
SK142W/8/HEM SK142W/10/HEM SK142W/7/HEM SK142W/11/HEM SK142W/9/HEM
89030-898CS 504.23 USD InStock
89030-898 89030-896 89030-894 89030-902 89030-900
SK Cleanroom Nitrile Gloves, Honeywell Safety
Gloves
Designed specifically for the cleanroom industry.
  • Beaded cuff
  • Embossed sizing
  • Specially manufactured cleanroom snap-tear packaging
  • Gloves are thicker for extra protection
  • Color: white
Made from a specially formulated nitrile latex material that produces 100% powder-free, case-hardened gloves. Provide outstanding solvent, puncture, and abrasion resistance. Hand-specific. Bisque finish for positive grip in wet and dry applications.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty