Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
VWR® Spinbar® Magnetic Stir Bars, Saturn
Supplier: Avantor
VWR® Spinbar®
37116-6003 37116-6002
89030-914CS 1036.54 USD InStock
89030-914 89030-912
VWR® Spinbar® Magnetic Stir Bars, Saturn
Stirrers Magnetic Stirring Bars
Many magnetic stirring bars can be impeded by powders in a liquid, but the Saturn Spinbar® Magnetic Stirring Bar rises to this stirring challenge.
  • Large sphere at the bar’s center lifts the stirring bar ends while spinning
  • Reduced surface contact area allows the bar to spin with decreased drag
  • Alnico magnet encapsulated in FDA Grade, USP Class VI Teflon® PTFE low-friction coating; durable and inert
  • Manufactured in a registered ISO 9001:2008 facility, as verified by SGS certification
  • Autoclavable at 121°C (250°F)
Magnetic stir bars are available with a length of 40 or 50mm (1.5 or 2") to match with a wide variety of vessels with both round and flat bottoms.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty