Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

VWR® Standard 1000 Orbital Shaker, 230 V 50 Hz, Export Only

Supplier: VWR
80863432
89032-090EA 1856.58 USD InStock
89032-090
VWR® Standard 1000 Orbital Shaker, 230 V 50 Hz, Export Only
Shakers and Mixers Orbital Shakers
The Standard Model 1000 shaker combines the size and economic benefits of a mini-shaker with the orbit and speed of a more traditional unit.

  • 230 V model
  • Variable speed microprocessor control
  • Low-profile design
  • Best-in-Class, 5-year warranty

The space-saving, low-profile design fits easily into most fume hoods and incubators.

Shakers are equipped with a ramping feature that slowly increases speed to the desired setpoint to avoid splashing or sample loss. Units feature a spill-resistant design that channels fluids away from internal components. Shakers can be run in cold rooms, incubators, and CO₂ environments from 0 to 40 °C (32 to 104 °F). Aluminum tray.

A cast aluminum base offers durability and added stability. The variable speed microprocessor control provides consistent uniform mixing action, and the powerful triple eccentric drive and brushless motor ensure reliable service in continuous duty. A basic speed knob with 1 to 10 dial settings controls all adjustments.

Certifications: RoHS certified. 230V unit is CE Marked.

Ordering information: Shaker is supplied with a nonskid rubber mat for securing Petri dishes and cell culture flasks. Unit also includes a 92” (234 cm) Euro type plug.

Caution: 230 V model.

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty