Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

VWR® Advanced 15000 Orbital Shakers, 230 V

Supplier: Avantor
VWR® Advanced
980327 980327-C 80863554 980325-C
89032-122EA 6482.17 USD InStock
89032-122 10027-164 89032-118 10027-160
VWR® Advanced 15000 Orbital Shakers, 230 V
Shakers and Mixers Orbital Shakers
Digital orbital shakers are designed for a wide range of applications such as solubility studies, extraction procedures, and cell cultures that require accurate and reproducible results.

  • Exclusive Accu-Drive™ shaking system
  • 150 lb. weight capacity
  • Best-in-Class, 5-year warranty

Touchpad controls with easy-to-read, independent LEDs for speed and time allow users to view all settings simultaneously

Distinctive features such as the exclusive Accu-Drive™ shaking system delivers exceptional speed control, accuracy, and durability. The system continuously monitors shaking speed and maintains set point even under changing loads. Displayed speed is accurate to 1% of setting. The variable speed microprocessor control provides consistent, uniform mixing action. Microprocessor wil display the last set point and will restart if power is interrupted. An integrated ramping feature slowly increases speed to the desired set point to avoid splashing or sample loss. The shaker's powerful triple eccentric drive and brushless motor provide reliable service in continuous duty and can handle loads up to 68 kg (150 lbs). 

Shakers can accommodate two stainless steel platform options. The universal platform is suitable for mounting flask clamps and test tube racks. The large vessel carrier features high side walls, and is ideal for use with carboys and large bottles. Flask clamps are available in either stainless steel or PVC styles. Media bottle clamps and microplate clamps are constructed of stainless steel. Pivoting test tube racks feature stainless steel construction. Standard test tube racks feature a PVC-coated steel construction. Both rack styles come with hardware for easy attachment to platforms. Aluminum tray.

The timer will display elapsed time or will shut off when programmed to a user defined limit. Displays will show last settings, even after power is turned off. A built-in load sensor detects unbalanced conditions and automatically reduces rpm to a safe speed, or stops unit operation entirely to protect samples. An audible alarm will sound when device is in timed mode. Shaker can run in cold rooms, incubators, and CO2 environments from -10 to 60°C (14 to 140°F). An RS-232 serial port provides two-way communication for data logging and unit control.

Certifications: RoHS certified. CE Marked, TUV listed.

Ordering information: Shakers are supplied with a nonskid rubber mat that fits the 61×91.4 cm (24×36") platform tray. Tray is suitable for holding Petri dishes and cell culture flasks. Unit also includes a 92” (234 cm) Euro type plug. All platform, clamp, and rack accessories must be ordered separately.

Order Now
 

ORDER

SPECIFICATIONS

ACCESSORIES

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty