Would you like to log in again?
DRUM LID PIG LATCHING BLACK
Supplier: NEW PIG CORP
Hinged, lockable Drum Lid used to provide easy access and security to materials stored in a 30-gallon open-head steel drum. Nitrile gasket completely seals lid to help maintain integrity of drum contents; helps reduce vapor emissions. Easy in tool-less in installation. Black.
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
DRUM LID PIG LATCHING BLACK 89032-424 DRM821-BK Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty