Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

VWR® Replica-Plating Tool

Supplier: VWR
VWR®
37848-6002 37848-6001 37848-6000
25395-382PK 178.35 USD InStock
25395-382 25395-380 89033-116
VWR® Replica-Plating Tool
Microplate and Petri Dish Replicators
The replica-plating tool utilizes a locking ring which secures sterile velveteen squares onto a PVC cylinder for easy control of the plating device.

  • For use with 90 to 100 mm Petri dishes
  • May be disinfected between uses with a brief rinse in 70% ethanol or chlorine bleach
  • Aluminum ring is 10.2 cm (4") diameter; PVC cylinder is 6.9 cm (23/4") high
  • Not autoclavable

To produce replicas, the Petri dish carrying microbial colonies is inverted and lowered onto the cloth surface, then lifted. A fresh sterile nutrient plate is then lowered onto the velveteen surface and lifted, leaving a replica of the original colonies in the new dish.

Ordering information: Tool comes with 12 sterile, white, 100% cotton, absorbent twill-backed velveteen squares; 15.2×15.2 cm (6×6"). Replacement velveteen squares are available; packed in a sealed bag with three inner locking bags each containing 12 squares, 36 pcs. total.

Order Now
 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty