Would you like to log in again?
Due to scheduled system maintenance, vwr.com will be temporarily unavailable from 6:00 am EST to 2:00 PM EST on Saturday, May 25. 
MegaMag Genie™ Magnetic Stirrers, Scientific Industries
Supplier: Scientific Industries
QuadMag Genie™
SI-2236 SI-3236
89037-468EA 1025.13 USD InStock
89037-468 89037-472
MegaMag Genie™ Magnetic Stirrers, Scientific Industries
Stirrers Magnetic Stirrers
Large volume magnetic stirrers are designed for large capacity applications
Powerful rare earth magnets automatically pull stir bars into position and stay coupled under the most demanding stirring conditions. The rugged ABS/polycarbonate housing and heavy-duty motor offer exceptional magnetic force and provide extended reliability. Stirrers feature non-skid feet to prevent the unit from moving during operation. The MegaMag Genie™ stirrers feature simple analog controls that allow user to easily change the desired speed. The digital MegaMag Genie™ stirrers feature true speed readout with feedback control that is adjustable in 10rpm increments. Digital unit also includes an adjustable timer. The analog and digital MegaMag Genie™ stirrers can stir up to 38L (10gal.)
Ordering information: The 230V units do not include a plug.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty