Would you like to log in again?
VWR® Casseroles, Porcelain
Supplier: VWR
VWR®
J-109 J-105 J-106 J-107 J-111 J-108
89037-970CS 686.24 USD InStock
89037-970 89037-972 89037-964 89037-966 89037-976 89037-968
VWR® Casseroles, Porcelain
Trays/Dishes/Pots Pots
Casseroles are constructed of chemical porcelain with pouring lip and flat handle.
  • Chemical porcelain construction
  • For use in high-temperature applications
  • Features pouring lip and flat handle
  • Glazed interior and exterior
Glazed inside and out, except for rim and top of handle. For use in temperatures up to 1150°C (2101°F).
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty