Would you like to log in again?
VWR® Combustion Boats, Porcelain
Supplier: Avantor
VWR®
J-115 J-114 J-113 J-112
89037-982CS 467.01 USD InStock
89037-982 89037-984 89037-978 89037-980
VWR® Combustion Boats, Porcelain
Boats Combustion Boats
Chemical-porcelain combustion boats feature a perforated handle and glazing inside and out, except for bottom surface.
  • Chemical porcelain construction
  • For use in high-temperature applications
  • Features pouring lip and flat handle
  • Glazed interior and exterior
For use in temperatures up to 1150°C (2102°F).
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty