Would you like to log in again?
VWR® Porous-Bottom Crucibles, Porcelain
Supplier: VWR
VWR®
J-149 J-150 J-148
89038-196CS 481.91 USD InStock
89038-196 89038-050 89038-048
VWR® Porous-Bottom Crucibles, Porcelain
Crucibles
Chemical-porcelain crucibles feature a porous bottom with a porous disc permanently formed into the walls.
  • Stable porosity
  • Consistent rate of flow
  • Chemical-porcelain construction
  • Cover is ordered separately.
Order Now
ORDER
ACCESSORIES
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty