Would you like to log in again?
VWR® Buchner Funnels, Table-Type
Supplier: VWR
VWR®
J-181 J-182
89038-114CS 725.4 USD InStock
89038-114 89038-112
VWR® Buchner Funnels, Table-Type
Funnels Filtration Funnels Büchner Funnels
These chemical-porcelain funnels are ideal for vacuum filtration and feature a slanting bottom that leads to a tubulated side outlet.
  • Perforated plate
  • Autoclavable
Designed with a wide mouth for convenience and a wide base that rests firmly on a tabletop for stability.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty