Would you like to log in again?
VWR® Buchner Filter Funnels
Supplier: VWR
VWR®
J-190 J-189 J-188 J-192 J-183 J-184 J-187 J-186 J-191 J-185
89038-132CS 590.03 USD InStock
89038-132 89038-120 89038-130 89038-118 89038-128 89038-116 89038-126 89038-124 89038-134 89038-122
VWR® Buchner Filter Funnels
Funnels Filtration Funnels Büchner Funnels
These chemical-porcelain Buchner filter funnels feature a fixed perforated plate and glazing inside and out, except for the rim.
  • Highly resistant to chemicals
  • Autoclavable
  • Glazed inside and out, except for the rim
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty