Would you like to log in again?
VWR® Hirsch Filter Funnels
Supplier: VWR
VWR®
J-194 J-193 J-195 J-196
89038-142CS 591.8 USD InStock
89038-142 89038-140 89038-138 89038-136
VWR® Hirsch Filter Funnels
Funnels Filtration Funnels Filter Funnels
These chemical-porcelain funnels feature a fixed perforated plate and glazing inside and out, except for rim.
  • Highly resistant to chemicals
  • Autoclavable
Filter funnels are available in a range of volume capacities and top outside diameters.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty