Would you like to log in again?
VWR® Mortars and Pestles, Porcelain
Supplier: VWR
VWR®
J-200 J-211 J-214 J-208 J-212 J-203 J-210 J-216 J-218 J-199 J-204 J-202 J-206 J-201 J-207 J-213 J-205 J-215 J-209 J-198
89038-176CS 661.3 USD InStock
89038-176 89038-154 89038-164 89038-152 89038-174 89038-184 89038-162 89038-172 89038-150 89038-160 89038-170 89038-180 89038-148 89038-158 89038-168 89038-146 89038-156 89038-178 89038-166 89038-144
VWR® Mortars and Pestles, Porcelain
Mortars and Pestles Mortars with Pestles
Mortars are constructed of chemical porcelain and feature a lip and a glazed exterior.
  • Porcelain construction
  • Glazed exterior, unglazed interior
  • Oversized pestle for comfort
Pestles are constructed entirely of chemical porcelain or with a porcelain grinding head and wooden handle. Wooden handle is not autoclavable. Pestles are also glazed to grinding surface and oversized for hand comfort.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty