Would you like to log in again?
Volumetric Flasks, Class A, Wide Mouth, Low Actinic, with PE Stopper, Chemglass
Supplier: Chemglass
CG-1620-50 CG-1620-100 CG-1620-05 CG-1620-500 CG-1620-200 CG-1620-1L CG-1620-250 CG-1620-20 CG-1620-10 CG-1620-2L CG-1620-25
89038-440EA 172.03 USD InStock
89038-440 89038-428 89038-438 89038-426 89038-436 89038-424 89038-434 89038-444 89038-432 89038-442 89038-430
Volumetric Flasks, Class A, Wide Mouth, Low Actinic, with PE Stopper, Chemglass
Flasks Volumetric Flasks
These volumetric flasks with wide mouth design facilitate mixing, pipette access, sampling and filling. The low actinic red stained glass provides protection to light sensitive solutions.
  • Clearly visible large permanent white letters
  • Heavy duty and heavy wall design
  • Allowing less than 1% transmission below 400 nm and 5% from 400 to 600 nm
The unique flat bottom interior permits the use of a standard stir bar.
Certifications: Flasks are calibrated ‘To Contain’ at Class A tolerances, per ASTM E288 specifications. Volumetric flask capacities of 20 ml, 500 ml, 1000 ml and 2000 ml are not included in the ASTM E288 specifications; however, flasks are calibrated to Class A tolerances.
Ordering information: Supplied complete with a PE stopper.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty