Would you like to log in again?
WYPALL® Microfiber Cloths with Microban® Protection, KIMBERLY-CLARK PROFESSIONAL®
Supplier: Kimberly-Clark
WYPALL*
83620 83610 83610 83630 83630 83620
89038-768CS 129.54 USD InStock
CA89038-764 89038-768 CA89038-766 89038-766 CA89038-768 89038-764
WYPALL® Microfiber Cloths with Microban® Protection, KIMBERLY-CLARK PROFESSIONAL®
Wipes Reusable Cleaning Cloths Cloths for General Applications
WYPALL® Microfiber Cloths with Microban® Protection are designed with the cleaning professional in mind.
When dampened with water, cloths physically remove up to 99% of bacteria from non-porous surfaces. The cloths are thick, heavy, durable, and remarkably absorbent, holding up to 8 times their weight in liquid for better and faster cleaning. They are a 'greener' cleaning option because of their ability to both clean and remove bacteria from surfaces without the use of added chemicals. Microban® Protection inhibits the growth of odor- and stain-causing bacteria on the cloth. Available in 3 different colors to reduce the chance of cross-contamination.
They are environmentally responsible, green-cleaning options that can be used without chemicals, thus reducing chemical costs. Because these cloths can be laundered 300 times, they reduce annual cleaning costs. Microfiber material is made by splitting fibers into tiny pieces and knitting them into a super-cleaning cloth that effectively traps soil and absorbs liquid for a streak-free shine.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty