Would you like to log in again?
DrainRack™ Jr. Microscope Slide Trays, Simport Scientific
Supplier: Simport
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
DrainRack™
M710-50G M710-50W M710-50W M710-50B M710-50G M710-50B M710-50P M710-50Y M710-50Y M710-50P
89039-728CS 96.56 USD InStock
CA48648-564 89039-728 89039-736 CA48648-572 89039-734 CA48648-570 CA89039-732 89039-732 89039-730 CA48648-566
DrainRack™ Jr. Microscope Slide Trays, Simport Scientific
Slide Holders
These rugged trays/drain racks can hold up to 100 microscope slides in 50 individual numbered slots
These microscope slide trays are made of plastic.
All slides are stored upright for easier insertion and removal. Simply tilt them forward or backward with one finger to easily pick up the desired slide. Bar codes can be read without having to remove the slides from the tray. Not autoclavable.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty