Would you like to log in again?
EP Scientific Septa, PTFE/Silicone, Thermo Scientific
Supplier: Thermo Scientific
S24-400-S2
89039-972CS 235.88 USD InStock
89039-972
EP Scientific Septa, PTFE/Silicone, Thermo Scientific
Septa
Standard flat disc PTFE/silicone septa for use with EP Scientific EPA Vials.
  • Fits the standard 24-414 open top cap
Certifications: Processed to meet EPA Performance Based Specifications for Volatile Organic Analysis.
Ordering information: Other sizes are available. Contact your Avantor sales representative for more information.
Order Now
ORDER
ACCESSORIES
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty