Would you like to log in again?
NorthFlex™ Light Task™ ESD Gloves, Honeywell Safety
Supplier: Honeywell Safety
NorthFlex™
NF15ESD/11XXL NF15ESD/9L NF15ESD/8M NF15ESD/10XL NF15ESD/7S NF15ESD/6XS
89040-172CS 83.84 USD InStock
89040-172 89040-182 89040-176 89040-174 89040-178 89040-180
NorthFlex™ Light Task™ ESD Gloves, Honeywell Safety
Gloves
Anti-Static conductive gloves with lightweight made of copper based Thunderon® with low surface resistivity.
  • Polyurethane coating on palm and fingertips
  • Applicable in manufacturing, municipal services and utilities
Certifications: Compliance with ANSI/ISEA 105 - ASTM D3389-05; ANSI/ISEA 105 - ASTM F1790-97; EN 388; EN 420 and OSHA Standard -29 CFR 1915.157.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty