Would you like to log in again?
Process2Clean® Liquid Cleaning Agents, Veltek Associates
Supplier: Veltek
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
Process2Clean®
PC-5-30G-02 PC-5-55G-02 PC-1-55G-02 PC-6-30G-01 PC-5-5G-02 PC-6-30G-02 PC-5-1G-02 PC-6-1G-01 PC-4-1G-01 PC-3-1G-02 PC-4-1G-02 PC-3-55G-02 PC-1-5G-02 PC-5-1G-01 PC-6-55G-01 PC-6-5G-02 PC-2-1G-02 PC-1-55G-01 PC-1-5G-01 PC-6-1G-02 PC-2-5G-02 PC-3-5G-01 PC-4-5G-02 PC-5-55G-01 PC-4-5G-01 PC-5-5G-01 PC-2-5G-01 PC-3-55G-01 PC-6-5G-01 PC-2-1G-01 PC-3-1G-01 PC-5-30G-01 PC-3-5G-02 PC-2-55G-01 PC-4-55G-01 PC-4-55G-02 PC-1-1G-01 PC-2-55G-02 PC-1-1G-02 PC-6-55G-02
89107-384EA 1678 USD InStock
89107-384 89040-326 89040-348 89107-386 89040-328 89107-388 89040-366 89040-388 89040-344 89040-322 89040-346 89040-368 89040-324 89040-384 89040-340 89040-362 89040-342 89040-364 89040-320 89040-386 89040-380 89040-382 89040-360 89107-390 89040-358 89040-336 89040-338 89040-376 89040-332 89040-354 89040-378 89040-356 89040-334 89040-350 89040-372 89040-374 89040-330 89040-352 89040-390 89040-370
Process2Clean® Liquid Cleaning Agents, Veltek Associates
Cleaning Agents Detergents and Surface Cleansers
These high-performance cleaning agents are specifically designed for critical clean-in-place applications and are engineered to effectively remove a multitude of product residues.
  • All products are formulated under the highest quality standards in a GMP manufacturing facility
  • Available in both sterile (filtered at 0.2µm and aseptically filtered) and nonsterile packaging
  • Sterile versions are ultra-clean and assure that less contamination is introduced to the system
  • Low-foaming
Process2Clean® 1 liquid alkaline agent is formulated with potassium hydroxide, surfactants, and other essential cleaning ingredients. Process2Clean® 2 liquid agent is an acidic based detergent. Both agents are capable of removing a wide array of proteins, excipients, fine chemicals, silicones, oils, petrolatum, polymers, and most types of organics.
Process2Clean® 3 hyodroxyacetic acid detergent is formulated with phosphoric acid, surfactants, and other essential cleaning ingredients. This agent is extremely effective in removing residues including a wide array of inorganic salts, scales, particulate carbon, urine scales (on polycarbonates, stainless steel, and other caging materials), proteins, excipients, fine chemicals, silicones, oils, petrolatum, polymers and most types of organics. Process2Clean® 4 and Process2Clean® 6 liquid agents are effective in cleaning ointments, creams, oils, waxes, greases, and petrolatum-based products. Both cleaning agents are designed for use in low-energy agitated immersion systems, pressure spray applications, or manual washing applications. Process2Clean® 5 liquid agent is designed specifically for use in recirculation and high-pressure cleaning applications. This agent is effective in removing a wide array of residues including oils, waxes, grease, and petrolatum.
Routine use of Process2Clean® 2 and 3 agents reduces corrosion, pitting, and rusting.
Ordering information: Agents include complete lot documentation package for traceability.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty