Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Isolator Tool™ and Covers, Micronova

Supplier: Micronova
Isolator Tool™
ITC11-911NS ITC620-911IR ITC6-9 ITC6-9NS ITC11911NSIR ITCAA-9IR ITC620-911 ITC11-9NS ITC11-9NSIR ITC6-911NS QPIT-09 QPIT-04 ITC6-911NSIR ITC6-9NSIR ITC6-9IR ITCAW-9 ITC6-4NS QDIT-09 ITC620-9IR ITC620-9 ITC11-9NSBB
76443-132CS 401.61 USD InStock
76443-132 76443-130 94023-776 89508-394 89040-914 76018-368 52428-594 94023-774 89040-926 89040-920 89092-312 76018-380 76018-370 89140-698 52428-592 76018-378 89089-356 76018-376 89402-988 76018-374 76018-372
Isolator Tool™ and Covers, Micronova
Mops
The Isolator Tool™ adapter with a 360° swivel works with a range of mop covers to efficiently clean and disinfect isolators, hoods, RABS, and small equipment in clean and aseptic areas.

  • Ergonomic cleaning for isolators, hoods, RABS, and other small equipment
  • Low-profile design and angled head adapter
  • Modular and extendable handles available
  • Available with stainless steel or plastic plates
  • For use with the SSU series Universal Handles or STAH series threaded handles
  • PolySorb™ Cover: 100% textured polyester for additional cleaning as well as sanitization
  • NovaPoly™ Cover: 100% smooth knit polyester matches traditional cleanroom wipe fabrics and provides excellent disinfectant dispersal
  • MegaTex® Cover: Polyamide, slightly abrasive yet absorbent
  • Microfiber Cover: 80% Polyester/20% Nylon for maximum absorbency and residue removal
  • Compatible with alcohol, strong disinfectants, and steam sterilization
  • Launderable for reuse
  • Autoclavable

The covers are available in a range of fabrics, and with an optional foam layer to hold disinfectant and facilitate wringing. Modular and extendable handles fit through small passes and allow extended reach. The all plastic version Isolator Tool™ and the mop covers are available gamma irradiated for aseptic areas. Designed for use with the SSU series Universal Handles or STA series threaded handles.

Ordering information: Fabric covers, adapters, and handles must be purchased separately. Slip covers are available irradiated; contact your Avantor sales representative for more information.

Order Now
 

ORDER

SPECIFICATIONS

ACCESSORIES

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty