Would you like to log in again?
Crimpers and Decappers for Microwave Vials, Chemglass
Supplier: Chemglass
CG-4930-20 CG-4930-21
89041-330EA 414.72 USD InStock
89041-330 89041-332
Crimpers and Decappers for Microwave Vials, Chemglass
Crimpers, Cappers and Decappers
Tools for crimping and decapping Aluminum seals on Microwave vials.
The crimper has an adjustment feature which will allow it to be used with a variety of septa, discs, stoppers or lined caps. The user may adjust the stroke or movement for each application as needed. The pliers decapper is an inexpensive way to remove seal caps by simply securing the seal between the jaws and squeezing the handles until cap pops off.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty