Would you like to log in again?
Adapters, Rotary Evaporator, Microwave Vials, PTFE, Chemglass
Supplier: Chemglass
CG-1318-A-10 CG-1318-A-12
89041-340EA 372.72 USD InStock
89041-340 89041-338
Adapters, Rotary Evaporator, Microwave Vials, PTFE, Chemglass
Adapters Adapters, Conical Ground Joints
Adapters feature a standard taper joint.
  • PTFE adapter allows convenient connection
  • Connecting microwave vial to a rotary evaporator bump
  • Trap having a standard taper joint size
Lower portion incorporates a threaded black Acetal compression cap that is slotted to accept the flange fitting on the vial.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty