Would you like to log in again?
AlphaTec® 02-100 Chemical resistant gloves, Ansell
Supplier: Ansell
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
AlphaTec®
988827 988825 902209 988826 0210011080 0210011110 0210011060 0210011070 988828 0210011090 988829
89041-858CS 1403.71 USD InStock
89041-858 CA89041-824 89041-856 CA89041-816 CA89041-814 89041-862 CA89041-818 89041-860 CA89041-820 89041-866 89041-864
AlphaTec® 02-100 Chemical resistant gloves, Ansell
Gloves
Style 02-100 flat-film gloves are constructed of a five-layer composite of polyethylene, nylon, and metallocene.
  • Can be worn beneath heavier gloves
  • Useful for handling in many different industries
  • Greater dexterity with winged thumbs
  • Hand-specific
The unique polymer and seam-fusion technology provide a secure seal against a wide range of chemicals, as well as a flexible, comfortable fit.
Uses include aircraft assembly; chemical processing, oil refining and petrochemicals; handling acids, alkalies, salts, ketones and alcohols; handling strong organic solvents; chemical plants and transportation; graphic arts and printing cleanup; handling specialty chemicals; and HazMat work. Can be worn as a liner under heavier gloves. Each glove is individually air-pressure tested and are made of HPPE.
Packaging: Gloves are packaged 12 pairs in a bag; 72 pairs in a carton.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty