Would you like to log in again?
Deck-/Panel-Mounted Pre-Rinse Units, WaterSaver Faucet
Supplier: WaterSaver Faucet
PR411-AOF PR1711WSA
89041-868EA 1226.4 USD InStock
89041-868 89041-870
Deck-/Panel-Mounted Pre-Rinse Units, WaterSaver Faucet
Faucets and Sinks Faucets
Deck- and panel-mounted pre-rinse units for hot and cold water feature a brass body with polished chrome-plated finish and forged brass four-arm handles with color-coded index disks.
  • Self-contained renewable valve units have replaceable stainless-steel seats
  • Integral check valves to prevent cross-mixing of hot and cold water
  • Fully assembled and factory tested prior to shipment
Inlet for panel-mounted unit is a 9.5 mm (3/8") NPT female union and is furnished with mounting shanks, locknuts, and washers. Inlet for deck-mounted unit is a 30.2 mm (13/16") male shank with two 9.5 mm O.D. (3/8") flexible copper tubes, locknut, washer, and inlet fittings. Pre-rinse assembly includes a self-closing squeeze valve with rubber-bound spray head, insulated handle, and locking ring. Valve is mounted on a spring-loaded 99.1 cm (39") stainless steel hose.
Order Now
ORDER
ACCESSORIES
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty