Would you like to log in again?
Whatman™ Roby Automated Filter Validation Kit, Whatman products (Cytiva)
Supplier: Whatman products (Cytiva)
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
Whatman™ Roby™
10463898 10463898
89042-104PK 963.3 USD InStock
89042-104 CA89042-104
Whatman™ Roby Automated Filter Validation Kit, Whatman products (Cytiva)
Filters Syringe Filters
Cytiva's Roby filter validation kit offers six different Roby automated syringe filters, along with a filter validation protocol and filter selection aid. Tubes of 25 filters are ready for loading into tablet tester systems.
  • At-a-glance information about the properties of each filter
  • Filter selection aid to select the ideal filter of the six available options
  • Optimized for SOTAX, Caliper (Zymark), Varian, and Erweka tablet tester systems
Whatman Roby syringe filters from Cytiva's business support automated filtering in tablet dissolution.
Automated tablet tester systems require automated filtration systems to support their process. The Roby filter validation kit from Cytiva offers multiple syringe filter options and tools to help select the optimal filter membrane for the particular dissolution being measured.
The included filter validation protocol ensures that the Roby syringe filter is correctly installed into the automated system being used.
Delivery information: The kit includes one tube of each membrane type. Syringe filters included: nylon, nylon with GF92 prefilter, regenerated cellulose, regenerated cellulose with GF92, glass fiber GF55, and glass fiber GF92. Includes filter validation protocol.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty