Would you like to log in again?
SP Bel-Art Spinbar® Capsule Magnetic Stirring Bar, Bel-Art Products, a part of SP
Supplier: Bel-Art
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
SCIENCEWARE®
F37152-0000
89042-248EA 53.88 USD InStock
89042-248
SP Bel-Art Spinbar® Capsule Magnetic Stirring Bar, Bel-Art Products, a part of SP
Stirrers Magnetic Stirring Bars
The Capsule Magnetic Stirring Bar is a polystyrene capsule containing a 30 x 8 mm (1.18 x 0.31") polygon magnetic stirring bar that spins freely inside the capsule while being prevented from spinning off center and ‘wandering’ on the base of a vessel.
  • Stirring bar contained to prevent damage to pH probes
  • Will not interfere with gels during staining procedures
  • Capsule can be easily disassembled for cleaning
  • Autoclavable
Containment of the stirring bar prevents it from impacting and damaging expensive pH probes during pH adjustments, and from interfering and damaging gels during staining/destaining procedures.
Large, flat capsule keeps the stirring bar consistently centered on the bottom of vertical electrophoresis units (e.g. Hoefer’s) to efficiently mix the running buffer.
Use of the capsule prevents the stirring bar from being impeded by chemicals which, in water, transform into a sticky gel (e.g. albumin) and enables efficient dissolving.
Manufactured in a registered ISO 9001:2008 facility, as verified by SGS certification.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty