Would you like to log in again?
E-Z Ex-Traction™ High Recovery Vials, Wheaton, DWK Life Sciences
Supplier: DWK Life Sciences
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
E-Z Ex-Traction™
W224618 W224634 W224632 W224601 W224630 W224619 W224603 W224613 W224617 W224636
89042-306CS 413.32 USD InStock
89042-306 89042-308 89042-296 89042-298 80076-564 89042-310 89042-300 89042-312 89042-302 89042-304
E-Z Ex-Traction™ High Recovery Vials, Wheaton, DWK Life Sciences
Vials
Precision-manufactured from low extractable borosilicate glass, these vials are ideal for compound management applications.
A variety of caps are available for these high recovery vials: black polypropylene open-top caps with fluoropolymer resin-faced silicone rubber liner, black phenolic screw caps with fluoropolymer resin-faced (14B) rubber liner, and closed and open-top white polypropylene screw caps with bonded fluoropolymer resin-faced silicone liners.
The conical well design ensures maximum recovery of high value samples. Vials feature a centered conical apex, which is important for automated applications. The large, smooth, and flat bottom provides stability as well as ample area for large bar codes.
Certifications: Vials meet ASTM Type I, Class A and USP Type I standards.
Order Now
ORDER
ACCESSORIES
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty