Would you like to log in again?
Dispensing Cannulas for SOCOREX® DOSYS™ Syringes, DWK Life Sciences
Supplier: DWK Life Sciences
851411 W860045 851412
53500-588EA 30.65 USD InStock
53500-588 53500-599 89044-546
Dispensing Cannulas for SOCOREX® DOSYS™ Syringes, DWK Life Sciences
Cannulae
Blunt end dispensing cannulas with Luer Lock. Ideal for use with 24 and 96 well microplates.
  • Stainless steel
  • Autoclavable
This unique dispensing system enables you to perform sample inoculations and cell feedings faster and more efficiently under sterile conditions.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty