Would you like to log in again?
Acura® Manual Pipettor Triopacks, DWK Life Sciences
Supplier: DWK Life Sciences
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
Acura®
W810323 W810327 W810320 W810324 W810322 W810326 W810321 W810325 W810319
89044-610CS 1266.82 USD InStock
89044-610 89044-608 89044-604 75800-380 89044-606 89044-600 89044-612 89044-602 89044-614
Acura® Manual Pipettor Triopacks, DWK Life Sciences
Pipettes Micropipettes
Each ready-to-use set includes three manual adjustable pipettors and accessories.
Nine different specific volume combinations are available with full coverage ranging from 0.1 µl to 10 ml.
Ordering information: Each set also includes tip samples, QC certificate, and operating instructions.
Order Now
ORDER
ACCESSORIES
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty