Would you like to log in again?
Pipettor Reagent Reservoirs, DWK Life Sciences
Supplier: DWK Life Sciences
Socorex™
W851364 W851239 W851368 W851365
89044-714EA 239.59 USD InStock
89044-714 89044-716 89044-710 89044-712
Pipettor Reagent Reservoirs, DWK Life Sciences
Reagent Reservoirs
Reagent reservoirs are designed for maximum recovery of liquid reagents.
  • Available in PP or PS
  • Sterile polystyrene reagent reservoirs are available in 8- or 12-channel formats
V-shaped 75 ml polypropylene reservoirs with open wells are well-suited for single-and multi-channel pipetting. The 125 ml reservoir includes a hinged lid.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty